Polymethylpentene High-Speed Funnels, Bel-Art
Fournisseur: Bel-Art
Certificats
A propos de cet article
For Non-Vacuum, High-Speed Filtration
- Internal ribs increase the filtration speed of filter paper
- Vertical outer ribs prevent airlock
- Unaffected by corrosive liquids
- Autoclavable; resists heat up to 177 °C
Customers Who Bought This Also Bought