Spécifications
- Description:Polymerase Chain Reaction Activity Kit
- Cat. no.:470149-852
- Supplier No.:BE-305
Spécifications
A propos de cet article
Teach Real-World Uses for PCR Technique
Demonstrate the various applications of polymerase chain reaction (PCR) technique as used in research, biotechnology and more. This hands-on kit contains everything necessary for amplification of a gene of interest, and can also be used to insert additional genetic information, such as tags, onto a particular gene. Materials provided for six groups of up to five students.
Demonstrate the various applications of polymerase chain reaction (PCR) technique as used in research, biotechnology and more. This hands-on kit contains everything necessary for amplification of a gene of interest, and can also be used to insert additional genetic information, such as tags, onto a particular gene. Materials provided for six groups of up to five students.