Polyhedral Alcohol Burner
# de catalogue 470020-698
Fournisseur: American Scientific
Small Business Enterprise
Spécifications
- Description:Polyhedralburner
- Cat. no.:470020-698
- Supplier No.:2066-11
Spécifications
A propos de cet article
Position the Flame at Just the Right Angle.
- Capacity: 125 mL
- Height: 4"
The polyhedral shape of this heavy-duty glass burner allows you to position the flame at just the right angle for different tasks. It comes with a nickel-plated snuffer cap and one wick. Uses 1/4" (6mm) thick replacement wicks.
Customers Who Bought This Also Bought