Science Kit Polyethylene Specimen Bags
Fournisseur: REGIONAL DISTRIBUTORS, INC.
Certificats
A propos de cet article
Find a multitude of uses for these bags in the laboratory and field.
- Bags can also be heat sealed
Made from clear, 'lay-flat' polyethylene, these bags resist moisture, acids, alkali, embalming fluids, and most solvents and preservatives.
Customers Who Bought This Also Bought