Pocket Test Thermometers
# de catalogue 470100-978
Fournisseur: Taylor Precision Products
Spécifications
- Description:Pocket Test Thermometer, 12.5 cm
- Cat. no.:470100-978
- Range:−10...+110 °C
- Supplier No.:6079J
Spécifications
A propos de cet article
Recalibratable thermometers feature a stainless steel thermistor and 2.5cm analog dial.
Thermometers measure from -10 to 110°C with 2° graduations. Includes a protective sheath with clip. Size: 12.5cm long.
Customers Who Bought This Also Bought