Plastic Spring Wave
# de catalogue 470039-692
Fournisseur: NADA SCIENTIFIC LTD.
Small Business Enterprise
Spécifications
- Description:Plastic Spring Wave
- Cat. no.:470039-692
- Supplier No.:C15-4103
Spécifications
A propos de cet article
Observe phase reversal at the fixed end of wave pulses with this spring used to test waves in fundamental and multiple vibrations.
- Ø×L: 2×50 cm
- Experiments by students
With many versatile applications, it can also be used in experiments to determine the speed of propagation of transverse and longitudinal waves. Lightweight, the non-tangling plastic spring is ideal for student use.
Customers Who Bought This Also Bought