Plastic Slide and Coverslip Lab Pack
# de catalogue 470148-952
Fournisseur: Raylabcon
Spécifications
- Description:Plastic slides and coverslip lab pack
- Cat. no.:470148-952
- Supplier No.:20205104
Spécifications
A propos de cet article
Start making your own slides!
- Explore slidemaking with this starter pack
- Hands-on learning
- Coverslips are made of clear PETG (polyethylene terephthalate glycol)
The set includes 50 slides 1×3", 0.5 mm thick and fifty 22 mm square coverslips and cleaning tissue.
Customers Who Bought This Also Bought