Plastic Knife Switches
# de catalogue 470005-872
Fournisseur: PRECISION MOULDED POLYMERS LIMITED
Spécifications
- Description:Plastic Knife Switch, Double Pole, Double Throw
- Cat. no.:470005-872
- Supplier No.:M30324
Spécifications
A propos de cet article
Virtually unbreakable, these sturdy knife switches feature brass poles and knives, making the switches well-suited for use in general-purpose classroom and laboratory experiments.
Customers Who Bought This Also Bought