Plastic Knife Switches
Fournisseur: PRECISION MOULDED POLYMERS LIMITED
Certificats
A propos de cet article
Plastic Knife Switches are easy to connect.
- Easy-To-Connect
- Unbreakable
- Teaching Tool for Open Circuit Lessons
- Rated at 5A
- Two Configurations Available
Basic components for any electrical lab, these switches are anchored onto plastic bases that allow visible connections for easy use. Educators use Plastic Knife Switches to explain open circuits.
These unbreakable switches are available in two configurations. They are rated at 5A.
Customers Who Bought This Also Bought