Plastic Cylinders
Supplier: Avantor
Certificates
About this item
These cylinders are made of Tenite® for moderate chemical resistance.
- Removable Square Base
- Handy Pour Spout
- Tenite and Plastic Construction
- For Use in Pre-High School Science Programs
- Easy-to-Read Single Graduations with Yellow Background
They are for use in pre-high school science programs.
Each cylinder has easy-to-read graduations and a convenient pour spout.
Customers Who Bought This Also Bought