Plastic Coverslips
# de catalogue 470148-658
Fournisseur: VWR International
Spécifications
- Numbering:#2
- Description:Plastic cover slips, rectangular
- Thickness:0.2 mm
- Cat. no.:470148-658
- Supplier No.:BI0095PK100
Spécifications
A propos de cet article
More Durable than Traditional Glass Coverslips for Younger Students
- Quality slide making supplies
- Primarily used for the younger science student
- Vinyl
The durable #2 thickness plastic is easier to work with, especially for temporary water mounts.
Remove lamination before use.
Customers Who Bought This Also Bought