ContactUsLinkComponent
Plastic Containers
Plastic Containers
# de catalogue 470206-268
Fournisseur:  U.S. FOODSERVICE, INC.
CONTAINER CLEAR POLYSTYRENE 32OZ PK125
Plastic Containers
# de catalogue 470206-268
Fournisseur:  U.S. FOODSERVICE, INC.
Spécifications
  • Description:
    32oz Polystyrene Container
  • Cat. no.:
    470206-268
  • Supplier No.:
    12504906
Spécifications
A propos de cet article
Polystyrene containers with snap-on lids for a variety of lab storage requirements.
  • Color: Clear
The 16 oz. container is sized at 5L×4W×2¹/₂"D.
The 32 oz. container is 6¹/₂L×4¹/₂W×2¹/₂"D.
Customers Who Bought This Also Bought