ContactUsLinkComponent
Plastic 'Pith' Ball Electroscope
Plastic 'Pith' Ball Electroscope
# de catalogue 470111-118
Fournisseur:  VWR International
PITH BALL ELECTROSCOPE PLASTIC
Plastic 'Pith' Ball Electroscope
# de catalogue 470111-118
Fournisseur:  VWR International
Spécifications
  • Description:
    Plastic “Pith” Ball Electroscope
  • Cat. no.:
    470111-118
  • Supplier No.:
    PH0894-SPL
Spécifications
A propos de cet article
A Cost-Effective Instrument
  • Durable
  • Quality
The plastic balls replace the more expensive pith balls and are slightly more durable but heavier. Balls are threaded and mount easily on the insulated stand. Use with friction rods or Van De Graaff generators.
Customers Who Bought This Also Bought