Ward's® Plant Mineral Deficiency Lab Activity
Catalog # 470021-146
Supplier: Avantor
Specifications
- Description:WARD’S Plant Mineral Deficiency Lab Activity
- Cat. no.:470021-146
- Supplier No.:WARD470021-146
Specifications
About this item
Combine Hydroponics and the Study of Mineral Deficiencies
With this WARD’S exclusive activity, your students germinate two types of seeds, then transfer them to various nutrient solutions for observation. Within two weeks, your students will become experts at identifying the symptoms of calcium, phosphorus, sulfur, magnesium, iron, nitrogen, and potassium soil deficiencies, all grown hydroponically. You will get a teacher’s manual and enough materials for ten setups.
Customers Who Bought This Also Bought