Plant Heating Mat
# de catalogue 470021-112
Fournisseur: HYDROFARM GARDEN CENTER SALES
Spécifications
- Description:Plant Heating Mat
- Cat. no.:470021-112
- Supplier No.:MT10006
Spécifications
A propos de cet article
Jump start your seeds with this heating mat that provides even, gentle warmth and encourages speedy germination for seeds and young plants.
The mat fits under common plant trays and warms the rooting area 10–20 degrees above the ambient temperature. The instructions for lighting, watering, and planting are printed on the mat.
20 W operation, 110 VAC. Size: 20"L x 10"W.
Customers Who Bought This Also Bought