Plant and Animal Cell Model Kit
Catalog # 470103-998
Supplier: Ward's Science
Specifications
- Description:Plant and Animal Cell Model Kit
- Cat. no.:470103-998
- Supplier No.:DD2030-KIT
Specifications
About this item
Using hema polymers, students create polymer cell models.
They then place their homemade organelles appropriately and cure the model with UV light to fix the components in place. The finished models offer students a constant visual aid of cellular structure and components. Includes materials for four models.
Customers Who Bought This Also Bought