Planck's Constant Determination Box
Catalog # 470018-432
Supplier: VWR International
Specifications
- Description:Planck's Constant Determination Box
- Cat. no.:470018-432
- Supplier No.:PH1365
Specifications
About this item
This box shows Planck's Constant in action.
- For Determining Approximate Value of Planck's Constant
- Seven LEDs from 470 nm to 940 nm
- Users Can Plot a Graph of Energy Input
This apparatus is designed to determine the approximate value of Planck's Constant using the elbow voltage in the characteristic curves of a number of LEDs. There are seven LEDs mounted on the box with sockets for voltage and current measurements. By monitoring the voltage at which each LED begins to emit light, users can plot a graph of energy input as a function of light emitted frequency and deduce an approximate value of Planck's Constant.
An external power supply for 9V is required. A 9V battery with suitable connecting cords can also be used.
Customers Who Bought This Also Bought