Planaria
# de catalogue 470001-778
Fournisseur: Avantor
Spécifications
- Size:1/4"
- Cat. no.:470001-778
- Packaging:Jar of 10
- Supplier No.:WARD470001-778
Spécifications
A propos de cet article
Formalin Preserved
Specimens of free-living turbellarian are relaxed and flattened to show structure.
Customers Who Bought This Also Bought