ContactUsLinkComponent
Graphite Coated Spheres
Graphite Coated Spheres
# de catalogue 470149-122
Fournisseur:  VWR International
GRAPHITE COATED BALL PK/3 3 CM D 5 G
Graphite Coated Spheres
# de catalogue 470149-122
Fournisseur:  VWR International
Spécifications
  • Description:
    Pith Balls Unthreaded
  • Cat. no.:
    470149-122
  • Supplier No.:
    PH45244A
Spécifications
A propos de cet article
Used to Detect Electrostatic Charge
Coated pith balls and spheres have a light conductive coating used in charge transfer experiments.
Customers Who Bought This Also Bought