ContactUsLinkComponent
Pig Uterus, Pregnant
Pig Uterus, Pregnant
# de catalogue 470000-606
Fournisseur:  Avantor
PIG UTERUS WITH 1-5 EMBRYOS PL/1 PM
Pig Uterus, Pregnant
# de catalogue 470000-606
Fournisseur:  Avantor
Spécifications
  • Description:
    Formalin Preserved Pig Uterus, Pregnant with Embryos
  • Size:
    1 to 5"
  • Cat. no.:
    470000-606
  • Packaging:
    Pail of 1
  • Supplier No.:
    WARD470000-606
Spécifications
A propos de cet article
Formalin preserved pig uterus. Available in pails.
  • Developemental biology
  • Reproductive structure
  • High quality preservation
These specimens are excellent supplements to our fetal pigs, because they allow students to see the pig’s initial developmental stages as well as the female reproductive system.
Customers Who Bought This Also Bought