ContactUsLinkComponent
Pig Brain with Eyes
Pig Brain with Eyes
# de catalogue 470000-920
Fournisseur:  Avantor
PIG BRAIN WITH EYES PAIL/1
Pig Brain with Eyes
# de catalogue 470000-920
Fournisseur:  Avantor
Spécifications
  • Cat. no.:
    470000-920
  • Packaging:
    Jar of 1
  • Supplier No.:
    WARD470000-920
Spécifications
A propos de cet article
Formalin preserved pig brain with eyes. Available in jars.
  • Mammalian anatomy
  • High quality preservation
Featuring intact eyes, dura mater, hypophysis, and some intact cranial nerves, this brain (which may lack the cerebellum) is ideal for demonstrations because it is slightly larger than the sheep brain.
Customers Who Bought This Also Bought