Piercing/Sucking Type, Squashbug Slide
# de catalogue 470181-622
Fournisseur: Avantor
Spécifications
- Description:Piercing/Sucking Type, Squashbug
- Cat. no.:470181-622
- Stain:unstained
- Supplier No.:WARD470181-622
- Orientation:Whole mount
Spécifications
A propos de cet article
Observe the Structure and Function Insect Mouthparts
- Identifying features clearly distinguishable
- Rigorous quality control standards
Squashbug demonstrating general body structures and piercing/sucking type mouthparts.
Customers Who Bought This Also Bought