Piercing Type, Mosquito, Whole Mount Slides
# de catalogue 470181-660
Fournisseur: Avantor
Spécifications
- Description:Piercing Type, Mosquito
- Cat. no.:470181-660
- Stain:unstained
- Supplier No.:WARD470181-660
- Orientation:Whole mount
Spécifications
A propos de cet article
Observed the Mouthpart of a Mosquito
- Identifying features clearly distinguishable
- Rigorous quality control standards
Demonstrates piercing type mouthparts of female mosquito. Unstained.
Customers Who Bought This Also Bought