Physical Pitch Sets
# de catalogue 470111-228
Fournisseur: Medvibe
Spécifications
- Description:Physical Pitch Set
- Cat. no.:470111-228
- Supplier No.:700-8STED
Spécifications
A propos de cet article
These Physical Pitch Sets are highly accurate.
- Accurate
- Clear Tone
- Eight in One Set
Each of the tuning forks in a set has a clear tone.
One Set of eight Tuning Forks consists of one each: 256C, 288D, 320E, 341.3F, 384G, 426.7A, 480B and 512C' .
Packaging: One set has eight tuning forks.
Customers Who Bought This Also Bought