Photographic Fog
# de catalogue 470024-590
Fournisseur: FORENSICS SOURCE
Spécifications
- Description:Photographic Fog
- Cat. no.:470024-590
- Supplier No.:PF-1
Spécifications
A propos de cet article
View the Trajectory of a Laser
For easier viewing and photographing of a laser’s trajectory, a few short bursts of photographic fog pointed at a laser beam make its entire length visible. 8 oz. can.
Customers Who Bought This Also Bought