Photographic Atlas
Fournisseur: FELINE PRESS
Certificats
A propos de cet article
James L. Castner
Designed specifically for college students in general entomology and insect taxonomy courses, this atlas features over 600 color photos that illustrate the structural characteristics and anatomical features of the major insect families and arthropod groups. In addition to the photos that cover approximately 190 different arthropod taxa, the guide also contains explanations and illustrations of external anatomy and insect development. Dichotomous keys to the family level for major orders and a glossary of specialized terms make this thorough reference a necessity for any entomologist.
Customers Who Bought This Also Bought