Photo Manual And Dissection Guides
Fournisseur: SQUARE ONE PUBLISHERS, INC
Certificats
A propos de cet article
Fred Bohensky
Perfect for comparative anatomy classes, these books all begin with a discussion of how humans compare to the specimen in question. They then cover basic dissection techniques, external features, and each individual system. Each chapter includes black and white photographs and self-tests to ensure that students are getting the most of out their dissections. Spiral bound.
Customers Who Bought This Also Bought