ContactUsLinkComponent
Ward's® Phenotype Expression Lab Activity
Ward's® Phenotype Expression Lab Activity
# de catalogue 470030-382
Fournisseur:  Avantor
PHENOTYPE EXPRESS KT MEETS AP REQ
Ward's® Phenotype Expression Lab Activity
# de catalogue 470030-382
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Phenotype Expression Kit
  • No. of Lab Groups:
    5
  • Cat. no.:
    470030-382
Spécifications
A propos de cet article
Investigate Transformation, a Basic Principle of Genetic Engineering
  • Genetically engineer non-pathogenic bacteria
  • Materials for 5 lab groups
  • Time Required: 90 minutes
In this experiment, the bacteria E. coli JM 101, which is sensitive to ampicillin and incapable of producing beta-galactosidase needed for lactose breakdown, is transformed by insertion of the pUC8 plasmid, receiving antibiotic resistance as well as the lac+ phenotype. In two 45-minute lab sessions, the traits are transferred, and the transformed cells are grown on agar plates containing the antibiotic and on plates with ampicillin and x-gal, a histochemical substrate for β; -galactosidase that yields a blue precipitate upon hydrolysis. Transformants appear as white colonies on plates with ampicillin and blue colonies on plates with ampicillin and x-gal. It comes with enough materials for five setups, as well as a teacher’s guide and student copymaster.
Materials required, not included: incubator.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Customers Who Bought This Also Bought