pH Test Papers
# de catalogue 470123-118
Fournisseur: Precision Laboratories
Spécifications
- Description:pH Test Papers, Litmus Red ACS (12 Vials)
- Cat. no.:470123-118
- Supplier No.:190-12V-100
Spécifications
A propos de cet article
Each stoppered vial contains 100 (1.8 x 2") strips.
Each stoppered vial contains 100 (1.8" x 2") strips.
Customers Who Bought This Also Bought