Permanent Ceramic Disc Magnet
Catalog # 470105-152
Supplier: Master Magnetics
Specifications
- Description:Permanent Ceramic Disc Magnet
- Cat. no.:470105-152
- Supplier No.:CD022800
Specifications
About this item
A permanent ceramic disc magnet with the North and South poles on opposite ends of the diameter.
58 mm diameter and 6.4 mm thick.
Customers Who Bought This Also Bought