Permanent Ceramic Block Magnet
# de catalogue 470149-774
Fournisseur: Master Magnetics
Spécifications
- Description:Permanent Ceramic Block Magnet
- Cat. no.:470149-774
- Supplier No.:CB60NMAG
Spécifications
A propos de cet article
A permanent ceramic block magnet with maximum magnetic efficiency, high inductance, and a minimum stray magnetic field.
The magnet is magnetized through the thickness to have complete freedom from pole orientation. Size: 50×25×10 mm.
WARNING: KEEP AWAY FROM ALL CHILDREN! This product contains (a) small magnet(s). Swallowed magnets can stick together across intestines causing serious infection and death. Seek immediate medical attention if magnet(s) are swallowed or inhaled.
Customers Who Bought This Also Bought