ContactUsLinkComponent
Peripheral Blood, Human Female Slide
Peripheral Blood, Human Female Slide
# de catalogue 470183-138
Fournisseur:  Avantor
PERIPHERAL BLOOD-HUMAN F (SM) GS NORMAL
Peripheral Blood, Human Female Slide
# de catalogue 470183-138
Fournisseur:  Avantor
Spécifications
  • Description:
    Peripheral Blood, Human Female
  • Cat. no.:
    470183-138
  • Stain:
    Giemsa
  • Supplier No.:
    WARD470183-138
  • Orientation:
    Smear
Spécifications
A propos de cet article
Note Female Blood Characteristics
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Normal. Barr bodies or drumsticks can be observed in at least 10% of the polymorphonuclear leucocytes.
Customers Who Bought This Also Bought