Peripheral Blood, Human Female Slide
# de catalogue 470183-138
Fournisseur: Avantor
Spécifications
- Description:Peripheral Blood, Human Female
- Cat. no.:470183-138
- Stain:Giemsa
- Supplier No.:WARD470183-138
- Orientation:Smear
Spécifications
A propos de cet article
Note Female Blood Characteristics
- Identifying features clearly distinguishable
- Rigorous quality control standards
Normal. Barr bodies or drumsticks can be observed in at least 10% of the polymorphonuclear leucocytes.
Customers Who Bought This Also Bought