Ward's® Peridinium
Fournisseur: Ward's Science
Certificats
A propos de cet article
Freshwater. Unicellular. Soil-water medium. Characterized by two flagella (one inside a groove at right angles to the other); most also have tests made of cellulose and are encrusted with silica.
Customers Who Bought This Also Bought