Peat Pots
# de catalogue 470147-452
Fournisseur: PARK SEED WHOLESALE
Spécifications
- Description:Peat Pots, 2 1/4"
- Cat. no.:470147-452
- Supplier No.:00624-CS-SP
Spécifications
A propos de cet article
Composed of peat moss and wood fiber, these pots allow the seedling to be planted without removing the pot. They allow easy penetration by growing roots and decompose as the plant develops.
Packaging: Package of 24.
Customers Who Bought This Also Bought