ContactUsLinkComponent
PCR-based VNTR Human DNA Typing Kit
PCR-based VNTR Human DNA Typing Kit
# de catalogue 470120-892
Fournisseur:  Edvotek
PCR VNTR HUMAN DNA TYPING KIT
PCR-based VNTR Human DNA Typing Kit
# de catalogue 470120-892
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    PCR-based VNTR Human DNA Typing Kit
  • Cat. no.:
    470120-892
  • Supplier No.:
    334-B
Spécifications
A propos de cet article
DNA polymorphisms are regions of DNA that vary greatly (are polymorphic) between individuals. These polymorphisms are frequently used to obtain a 'DNA fingerprint of individuals.
One polymorphic DNA region, known as the D1S80 locus variably contains between 14 and 40 copies of a 16 base pair repeat; this is an example of a VNTR (variable number of tandem repeats) polymorphism.
In this experiment, students isolate their DNA from cheek cells and perform PCR to fingerprint their DNA at the D1S80 locus, using technology used by FBI laboratories in a safe classroom experiment. Students can also quantitatively measure the size of PCR products to determine individual fingerprints. Safe experimental design enables students to perform the experiment using hair or sputum. Includes instructions and materials for 25 reactions.
Requires electrophoresis equipment, thermal cycler, white light or UV transilluminator, micropipets and microcentrifuge, all available separately.
Customers Who Bought This Also Bought