PCR-Based DNA Fingerprinting Kit
# de catalogue 470120-896
Fournisseur: Edvotek
Spécifications
- Description:PCR-Based DNA Fingerprinting Kit
- No. of Lab Groups:5
- Cat. no.:470120-896
- Supplier No.:371
Spécifications
A propos de cet article
Students explore Forensic Science with the PCR-Based DNA Fingerprinting Kit.
- Lab Encourages Independent Inquiry
- Time Required: 75 Minutes plus Two Hours for PCR
- Students Practice PCR in the Classroom
- Instructions Included
- PCR EdvoBeads Included
This kit provides easy-to-follow instructions, so students can develop various crime scene scenarios independently.
Plasmid DNA, when amplified by PCR, provides materials that represent individual DNA profiles. Students use these outcomes to solve a crime.
Ordering information: This kit includes instructions, PCR EdvoBeads, DNA templates, primers, DNA ladder, ultrapure water, wax beads, agarose, loading dye, electrophoresis buffer, InstaStain Ethidium Bromide and FlashBlue stain. Micropipettes that measure between 5 and 50 µl, tips, a thermal cycler, electrophoresis apparatus, a power supply, a microwave or hot plate, and a UV transilluminator are required but not included.
Packaging: Some components require freezer storage on arrival.
Customers Who Bought This Also Bought