ContactUsLinkComponent
Polymerase Chain Reaction (PCR) Simulation Kit
Polymerase Chain Reaction (PCR) Simulation Kit
# de catalogue 470039-554
Fournisseur:  Edvotek
AMPLIFICATION OF DNA BY PCR
Polymerase Chain Reaction (PCR) Simulation Kit
# de catalogue 470039-554
Fournisseur:  Edvotek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    PCR- Polymerase Chain Reaction Simulation Kit
  • Cat. no.:
    470039-554
  • Supplier No.:
    103
Spécifications
A propos de cet article
Ideal for Beginning PCR Instruction
This easy-to-use simulation activity introduces students to the indispensable techniques, principles, and applications of the Polymerase Chain Reaction (PCR) without the need for a thermal cycler. Students will develop the skills needed for further application and study of PCR. Ready-to-Load™ DNA samples are included to allow for quick and easy setup. Kit also includes UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, InstaStain® Blue and FlashBlue™ stain, calibrated pipette, 100 ml graduated cylinder, and microtipped transfer pipettes. Activity requires an electrophoresis apparatus, power supply, automatic micropipette and tips, balance, microwave or hot plate, and visualization system. Material supplied for six gels. Time requirements: 60 minutes.
Customers Who Bought This Also Bought