Order Entry
ContactUsLinkComponent
Paper Binoculars
Paper Binoculars
# de catalogue 470102-468
Fournisseur:  Ajax Scientific
BINOCULARS PAPER PKG/10
Paper Binoculars
# de catalogue 470102-468
Fournisseur:  Ajax Scientific

Spécifications

  • Magnification:
  • Description:
    Paper Binoculars
  • Cat. no.:
    470102-468
  • Supplier No.:
    32-501

Spécifications

A propos de cet article

These folding, pocket-sized binoculars feature 25mm lenses providing 3X magnification.

Excellent for introducing young students to the uses of binoculars and for field trips and animal observation, they are inexpensive enough that every student can have one.

Size: 3 x 25 cm.

Packaging: Package of 10.
Customers Who Bought This Also Bought