Pails with Steel Handles
# de catalogue 470144-736
Fournisseur: Paragon Manufacturing
Spécifications
- Capacity:13 L
- Color:White
- Description:Pails with steel handles, pail
- Cat. no.:470144-736
- Supplier No.:3W
Spécifications
A propos de cet article
Molded of polyethylene with slightly tapered sides and plastic grip. Easily stacked for storage. Lid sold separately.
Customers Who Bought This Also Bought