Ward's® Live Oscillatoria Culture
Fournisseur: Ward's Science
Certificats
A propos de cet article
Oscillatoria is a genus of filamentous green algae that moves by sliding filaments back and forth against each other.
- Filamentous, Freshwater green algae
- Unbranched trichome, highly sheathed
- Genus gets its name from the oscillating motion it exhibits
Genus: Oscillatoria Species: not specified
Customers Who Bought This Also Bought