Optic Nerve Slide
# de catalogue 470182-736
Fournisseur: Avantor
Spécifications
- Description:Optic Nerve
- Cat. no.:470182-736
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-736
- Orientation:Cross section
Spécifications
A propos de cet article
Transmits all Visual Information to the Brain
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Bundles of nerve fibers surrounded by sheath layers.
Customers Who Bought This Also Bought