Opossum Skull, Ward’s®
# de catalogue 470028-022
Fournisseur: Avantor
Spécifications
- Description:Opossum Skull
- Height:2"
- Cat. no.:470028-022
- Length:4 .5"
- Width:2.5"
- Supplier No.:WARD470028-022
- No. of Parts:1
Spécifications
A propos de cet article
This unmounted skull of the common North American Didelphis virginiana shows the extra teeth, which is a characteristic of marsupials.
- Natural bone
- Professionally cleaned and prepared
Ordering information: This is a real specimen, so size and appearance may vary from the image.
Customers Who Bought This Also Bought