One Bite Out of Crime Kit
Fournisseur: Kemtec
Certificats
A propos de cet article
Learn the Principles of Forensic Odontology
- Personalized Jaw Impressions
- Easy to Use Dental Charts
Teeth not only provide a unique identifier for each person, they also last longer after death than any other physical element. Using the information provided, students will make a cast of their own upper jaw, learn to identify teeth placement, make dental charts, learn the metric analysis of a bite mark, and match dental charts with an unidentified body to provide identification. The kit includes enough materials for six setups, as well as instructor, student, and assessment manuals.
Customers Who Bought This Also Bought