Object and Marker
# de catalogue 470006-938
Fournisseur: ELECTRO MECHANICAL INSTRUMENTS
Spécifications
- Description:Object and Marker
- Cat. no.:470006-938
- Supplier No.:6010
Spécifications
A propos de cet article
Perfect for Your Meter Stick Optics Bench
Used as an image for lens and mirror experiments on a meter stick optical bench.
Customers Who Bought This Also Bought