Nylon Semimicro Test Tube Brush
Fournisseur: Mill-Rose
Certificats
A propos de cet article
For test tubes 5 to 11 mm in diameter. Total length: 14.6 cm; length of fiber section: 7 cm; diameter: 1.3 cm.
Customers Who Bought This Also Bought
For test tubes 5 to 11 mm in diameter. Total length: 14.6 cm; length of fiber section: 7 cm; diameter: 1.3 cm.