Nutrient Agar
# de catalogue 470193-230
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Description:Dehydrated Nutrient Agar
- Cat. no.:470193-230
- Supplier No.:880004
Spécifications
A propos de cet article
Multipurpose media. Supports the growth of a variety of microorganisms.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 tubes.
Customers Who Bought This Also Bought