Newton's Hovercraft
Fournisseur: INDUSTRIAL SUPPORT INC.
Certificats
A propos de cet article
Students discover the joy of Newton’s Laws with Newton's Hovercraft.
- Hovercraft Width: 40"
- No Air Track Needed
- Fun Kinematics Lesson
- Large-Scale Action and Reaction Demonstration
- Students Observe Newton's Laws in Action
They can ride on a cushion of air and experience near-frictionless motion. The 40" wide hovercraft is easy to use, and it doesn't need an air track.
Now teachers can demonstrate action and reaction on a large scale.
Ordering information: This kit includes a hovercraft and instructions for lab activities. A leaf blower, used to operate the hovercraft, is sold separately.
Customers Who Bought This Also Bought