ContactUsLinkComponent
Ward's® New York Vikings: A Lab Activity in Forensic Anthropology and Geology
Ward's® New York Vikings: A Lab Activity in Forensic Anthropology and Geology
# de catalogue 470125-674
Fournisseur:  Avantor
WARD S NY VIKINGS: A FORENSIC LAB ACT
Ward's® New York Vikings: A Lab Activity in Forensic Anthropology and Geology
# de catalogue 470125-674
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    WARD'S New York Vikings: A Lab Activity in Forensic Anthropology and Geology
  • No. of Lab Groups:
    3
  • Cat. no.:
    470125-674
  • Supplier No.:
    WARD470125-674
Spécifications
A propos de cet article
Analyze human bone, artifacts, and rocks.
  • Grades 9-12
  • Real World Case Based Scenario
  • Estimated class/lab time required: 180 minutes over 4 days
In this forensic science activity, students will apply hands-on forensic techniques to analyze a scenario involving multiple lines of anthropological, archaeological, and geological evidence from human bone replicas, artifact photos, and rocks. A human skeleton found in the woods in northern New York and associated artifacts and rocks suggest a possible Viking burial. But is it? Or is it a creative cover-up for a murder scene? Three student teams work together to analyze the evidence, reconstruct the scene, and determine if a crime was committed and, if so, the identity of the perpetrator. This detailed and fascinating study of bones and stones was developed in collaboration with Tyler G. O’Brien, Ph. D., Associate Professor of Anthropology at the University of Northern Iowa, and Neal R. O-Brien, Emeritus Professor of Geology at SUNY at Potsdam, for students in high school and early college. The activity can be completed in four 45-minute lab sessions with additional research time outside of class.
Materials include:
Replicas of three adult human bones (skull, femur and pelvis)
Metric vernier caliper
Caliper
Set of six rocks
Limestone
Magnifiers
Pipettes
Hydrochloric acid
Literature
Also needed are a computer with Internet access
Atlas of the human skeleton
World map
Customers Who Bought This Also Bought