Neutralization by the Drop Titration Method Kit
# de catalogue 470200-526
Fournisseur: Kemtec
Spécifications
- Description:Neutralization by the Drop Titration Method Kit
- No. of Lab Groups:6
- Cat. no.:470200-526
- Includes:Teacher ‘s Guide and Student Copymasters, Special Needs Reference Sheet
- Supplier No.:4-114
Spécifications
A propos de cet article
The Neutralization by the Drop Titration Method Kit contains an easy and hands-on titration experiment.
- Ideal for Grades 8-12
- Easy-To-Use
- Good Fit for Students with Limited Lab Background
- No Preparation Required
- Necessary Materials Included
It's a great way to teach the principles of acid and base titration to students with a limited laboratory background. It eliminates the need for expensive laboratory equipment for teaching titration and includes all necessary materials, alleviating extensive lab preparation.
The kit is ideal for leading discussions about titrations, acids, bases, pH, buffers, or environmental or physiological buffering systems in a range of science applications from biology to environmental sciences to physiology.
Ordering information: This kit includes a SDS universal indicator, sodium hydroxide, hydrochloric acid, phenolphthalein indicator, dropper bottles, toothpicks, a universal indicator chart, and depression plates.
Customers Who Bought This Also Bought