Nerve and Muscle Action Microslide
Fournisseur: SUPERTEK SCIENTIFIC LLC
Certificats
A propos de cet article
Students investigate nervous and muscle tissues of the body.
- Arrows Help Students Locate Important Features
- Mounted in a Clear Plastic Holder That Protects It on Both Sides
- Set of 15 Includes Detailed Lesson Plan on the Topic Under Study
- Durable, Inexpensive Teaching Resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions.
You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
Ordering information: This set includes single cell (100x); synapse (100x/em, 300,000x); axons (cs, 400x/ls, 200x); myelinated axons (600x); nerve-muscle junction (400x); synaptic knobs (em, 300,000x); striated muscle cell (1000x); sarcomeres, relaxed (em, 45,000x); sarcomeres, constructed (em, 31,000x).
Customers Who Bought This Also Bought